Assay Details
Target Gene Details
Entrez Gene ID: | 64058 |
Gene Name: | PERP, TP53 apoptosis effector |
Gene Aliases: |
1110017A08Rik, KCP1, KRTCAP1, PIGPC1, THW |
Location: |
Chr.10:18845071-18857073 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Perp | NM_022032.4 | 1 | 285 | NP_071315.1 |
AF249870.1 | 1 | 286 | AAF64306.1 | |
AK003734.1 | 1 | 226 | BAB22968.1 | |
AK009184.1 | 1 | 223 | BAB26126.1 | |
AK009190.1 | 1 | 224 | BAB26130.1 | |
BC021772.1 | 1 | 210 | AAH21772.1 |