Assay Details
Target Gene Details
Entrez Gene ID: | 215819 |
Gene Name: | NHS-like 1 |
Gene Aliases: |
5730409E15Rik, A630035H13Rik, A730096F01, BC013565, D10Bwg0940e, mKIAA1357 |
Location: |
Chr.10:18318558-18533892 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nhsl1 | NM_173390.3 | 1 | 145 | NP_775566.3 |
AK017530.1 | 1 | 146 | BAB30793.1 | |
AK038892.1 | 1 | 121 | BAC30160.2 | |
AK041760.1 |