Assay Details
Target Gene Details
Entrez Gene ID: | 18029 |
Gene Name: | nuclear factor I/C |
Gene Aliases: |
1110019L22Rik, 1500041O16Rik, AA589446, AI746521, NF1-C |
Location: |
Chr.10:81396191-81431167 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nfic | NM_008688.3 | 10 | 3856 | NP_032714.1 |
NM_026756.2 | 9 | 3779 | NP_081032.2 | |
AK147496.1 | 9 | 3764 | BAE27951.1 |
More Information
Set Membership: |
Intragenic Exonic |