Assay Details
Target Gene Details
Entrez Gene ID: | 16798 |
Gene Name: | large tumor suppressor |
Gene Aliases: |
AW208599 |
Location: |
Chr.10:7681209-7716461 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lats1 | NM_010690.1 | 2 | 560 | NP_034820.1 |
AK038612.1 | 2 | 561 | BAC30063.1 | |
BC158092.1 | 1 | 406 | ||
BC158123.1 | 2 | 555 |
More Information
Set Membership: |
Intragenic Exonic |