Assay Details
Target Gene Details
Entrez Gene ID: | 215814 |
Gene Name: | coiled-coil domain containing 28A |
Gene Aliases: |
1700009P13Rik, AI480677 |
Location: |
Chr.10:18213685-18234991 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc28a | NM_144820.3 | 6 | 920 | NP_659069.2 |
AK148447.1 | 6 | 921 | BAE28559.1 | |
BC027759.1 | 6 | 928 |
More Information
Set Membership: |
Intragenic Exonic |