Assay Details
Target Gene Details
Entrez Gene ID: | 17937 |
Gene Name: | Ngfi-A binding protein 2 |
Gene Aliases: |
AI451907 |
Location: |
Chr.10:127660918-127666703 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nab2 | NM_001122895.1 | 2 | 623 | NP_001116367.1 |
NM_008668.2 | 2 | 623 | NP_032694.2 | |
AK155501.1 | 2 | 521 | BAE33295.1 | |
AK158688.1 | 2 | 553 | BAE34612.1 | |
AK171180.1 | 2 | 503 | BAE42297.1 | |
BC045139.1 | 2 | 507 | AAH45139.1 | |
U47543.1 | 2 | 598 | AAC52650.1 |