Assay Details
Target Gene Details
Entrez Gene ID: | 14012 |
Gene Name: | myelin protein zero-like 2 |
Gene Aliases: |
Eva, Eva1 |
Location: |
Chr.9:45040291-45054043 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mpzl2 | NM_007962.4 | 6 | 2492 | NP_031988.2 |
AK032788.1 | 6 | 2273 | BAC28022.1 | |
AK040812.1 | 2 | 1460 | ||
AK166341.1 | 6 | 2223 | BAE38716.1 | |
AK167263.1 | 6 | 2256 | BAE39377.1 | |
AK170816.1 | 6 | 2258 | BAE42048.1 | |
BC015076.1 | 6 | 2318 | AAH15076.1 |
More Information
Set Membership: |
Intragenic Exonic |