Assay Details
Target Gene Details
Entrez Gene ID: | 14012 |
Gene Name: | myelin protein zero-like 2 |
Gene Aliases: |
Eva, Eva1 |
Location: |
Chr.9:45040291-45054043 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mpzl2 | NM_007962.4 | 1 | 75 | NP_031988.2 |
BY230064.1 | 1 | 75 |
More Information
Set Membership: |
Intragenic Exonic |