Assay Details
Target Gene Details
Entrez Gene ID: | 22040 |
Gene Name: | three prime repair exonuclease 1 |
Gene Aliases: |
1661, AU041952 |
Location: |
Chr.9:109057932-109059723 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trex1 | NM_001012236.1 | 2 | 889 | NP_001012236.1 |
NM_011637.6 | 2 | 903 | NP_035767.4 | |
AF151106.3 | 2 | 889 | AAD48775.2 | |
AF319574.1 | 1 | 1604 | AAK07621.1 | |
AK009899.1 | 2 | 890 | BAB26571.2 | |
AK087832.1 | 2 | 890 | BAC40020.1 | |
AK171916.1 | 2 | 878 | BAE42731.1 | |
AU041952.1 | 1 | 172 | ||
BC011133.1 | 2 | 910 | AAH11133.1 | |
CO041929.1 | 1 | 184 |