Assay Details
Target Gene Details
Entrez Gene ID: | 16000 |
Gene Name: | insulin-like growth factor 1 |
Gene Aliases: |
C730016P09Rik, Igf-1, Igf-I |
Location: |
Chr.10:87858270-87937047 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igf1 | NM_001111274.1 | 5 | 5973 | NP_001104744.1 |
NM_001111275.1 | 4 | 6003 | NP_001104745.1 | |
NM_001111276.1 | 4 | 5921 | NP_001104746.1 | |
NM_010512.4 | 5 | 6055 | NP_034642.2 |
More Information
Set Membership: |
Intragenic Exonic |