Assay Details
Target Gene Details
Entrez Gene ID: | 319415 |
Gene Name: | heparan sulfate (glucosamine) 3-O-sulfotransferase 5 |
Gene Aliases: |
D930005L05Rik, Gm1151, Hs3ost5 |
Location: |
Chr.10:36506804-36834397 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hs3st5 | NM_001081208.2 | 1 | 541 | NP_001074677.1 |
NM_001253355.1 | 1 | 541 | NP_001240284.1 | |
NM_001253356.1 | 1 | 541 | NP_001240285.1 | |
AK031910.1 | 1 | 531 | BAC27601.1 | |
AK086098.1 | 1 | 460 | ||
BC147490.1 | 1 | 534 | ||
BC147493.1 | 1 | 534 | ||
BC150973.1 | 1 | 534 | ||
BC158004.1 | 1 | 534 | ||
CJ113442.1 | 1 | 542 |
More Information
Set Membership: |
Intragenic Exonic |