Assay Details
Target Gene Details
Entrez Gene ID: | 54427 |
Gene Name: | DNA (cytosine-5-)-methyltransferase 3-like |
Gene Aliases: |
D6Ertd14e, ecat7 |
Location: |
Chr.10:78042287-78063622 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dnmt3l | NM_001081695.2 | 14 | 1669 | NP_001075164.1 |
NM_001284197.1 | 12 | 1510 | NP_001271126.1 | |
NM_001284198.1 | 6 | 1306 | NP_001271127.1 | |
NM_001284199.1 | 6 | 1133 | NP_001271128.1 | |
NM_001284200.1 | 5 | 967 | NP_001271129.1 | |
NM_019448.4 | 13 | 1523 | NP_062321.1 | |
AF220524.1 | 12 | 1454 | AAF73868.1 | |
AJ404467.1 | 12 | 1495 | CAB94726.1 | |
AK010434.1 | 13 | 1524 | BAB26936.1 | |
AV511576.1 | 1 | 98 | ||
BC083147.1 | 12 | 1508 | AAH83147.1 | |
EF051621.1 | 6 | 1306 | ||
EF051622.1 | 6 | 1133 | ||
EF051623.1 | 5 | 967 | ||
EF051624.1 | 14 | 1669 |
More Information
Set Membership: |
Intragenic Exonic |