Assay Details
Target Gene Details
Entrez Gene ID: | 20397 |
Gene Name: | sphingosine phosphate lyase 1 |
Gene Aliases: |
AI428538, D10Xrf456, S1PL, Spl |
Location: |
Chr.10:61098642-61147686 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sgpl1 | NM_009163.3 | 15 | 3337 | NP_033189.2 |
AK036747.1 | 15 | 3225 | BAC29562.1 | |
AK037789.1 | 15 | 3222 | BAC29872.1 | |
AK043024.1 | 15 | 3224 | BAC31437.1 | |
AK049342.1 | 15 | 3245 | BAC33695.1 | |
AK075851.1 | 6 | 2323 | ||
AK150053.1 | 15 | 3232 | BAE29269.1 | |
AK173137.1 | 15 | 3266 | BAD32415.1 | |
AK191088.1 | ||||
AK192177.1 | 1 | 339 | ||
AK193544.1 | 1 | 339 | ||
AK196336.1 | ||||
BC026135.1 | 15 | 3283 | AAH26135.1 |