Assay Details
Target Gene Details
Entrez Gene ID: | 20393 |
Gene Name: | serum/glucocorticoid regulated kinase 1 |
Gene Aliases: |
Sgk |
Location: |
Chr.10:21882184-21999903 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sgk1 | NM_001161845.2 | 14 | 2885 | NP_001155317.2 |
NM_001161847.2 | 12 | 2386 | NP_001155319.1 | |
NM_001161848.2 | 12 | 2179 | NP_001155320.1 | |
NM_001161849.2 | 12 | 2260 | NP_001155321.1 | |
NM_001161850.2 | 12 | 2367 | NP_001155322.1 | |
NM_011361.3 | 12 | 2235 | NP_035491.1 | |
AF139638.1 | 12 | 2219 | AAD43302.1 | |
AF205855.1 | 12 | 2216 | AAF19429.1 | |
AK086892.1 | 12 | 2219 | ||
AK132234.1 | 12 | 2307 | BAE21048.1 | |
AK146037.1 | 12 | 2219 | BAE26849.1 | |
AK146062.1 | 12 | 2216 | BAE26871.1 | |
AK159131.1 | 12 | 2219 | BAE34843.1 | |
AK167364.1 | 12 | 2216 | BAE39461.1 | |
BC002222.1 | 5 | 1413 | AAH02222.1 | |
BC005720.1 | 12 | 2196 | AAH05720.1 | |
BC070401.1 | 14 | 2885 | AAH70401.1 | |
BY493368.1 | 1 | 283 |
More Information
Set Membership: |
Intragenic Exonic |