Assay Details
Target Gene Details
Entrez Gene ID: | 244954 |
Gene Name: | protease, serine 35 |
Gene Aliases: |
6030424L22Rik, P3D9 |
Location: |
Chr.9:86743240-86758443 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss35 | NM_178738.3 | 2 | 953 | NP_848853.2 |
AK030671.1 | 2 | 938 | BAC27073.1 | |
AK031411.1 | 2 | 938 | BAC27392.1 | |
AK031644.1 | 2 | 938 | BAC27491.1 | |
AK076281.1 | 2 | 938 | ||
BC075675.1 | 2 | 922 | AAH75675.1 | |
DQ223037.1 | 2 | 913 |
More Information
Set Membership: |
Intragenic Exonic |