Assay Details
Target Gene Details
Entrez Gene ID: | 19263 |
Gene Name: | protein tyrosine phosphatase, receptor type, B |
Gene Aliases: |
3230402H02Rik, C130094E24, Ptpz, Rptpb, VE-PTP, Veptp |
Location: |
Chr.10:116275516-116389538 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprb | NM_029928.2 | 4 | 1024 | NP_084204.2 |
AK019450.1 | 6 | 1687 | BAB31726.1 | |
AK087168.1 | 1 | 1154 | ||
AK147439.1 | 4 | 1026 | BAE27912.1 | |
AK147668.1 | 4 | 1026 | BAE28060.1 | |
AY077755.1 | 4 | 902 | AAL75813.1 | |
BC141006.1 | 4 | 820 | ||
BC145111.1 | 4 | 820 |
More Information
Set Membership: |
Intragenic Exonic |