Assay Details
Target Gene Details
Entrez Gene ID: | 171580 |
Gene Name: | microtubule associated monooxygenase, calponin and LIM domain containing 1 |
Gene Aliases: |
MICAL, MICAL-1, Nical |
Location: |
Chr.10:41475860-41487030 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mical1 | NM_001164433.1 | 23 | 3248 | NP_001157905.1 |
NM_138315.2 | 24 | 3467 | NP_612188.1 | |
AK154665.1 | 23 | 3313 | ||
AK154930.1 | 24 | 3450 | ||
AK157196.1 | 24 | 3404 | ||
AK159369.1 | 23 | 3195 | BAE35027.1 | |
AK170382.1 | 24 | 3409 | ||
AK170827.1 | 24 | 3439 | ||
AK170874.1 | 24 | 3449 | ||
AK170881.1 | 24 | 3433 | BAE42090.1 | |
AK171234.1 | 24 | 3434 | BAE42330.1 | |
AK171429.1 | 24 | 3446 | BAE42447.1 | |
AK171479.1 | 24 | 3440 | BAE42481.1 | |
AK172562.1 | 24 | 3442 | ||
AK172716.1 | 24 | 3394 | ||
AK188198.1 | 1 | 103 | ||
BC021477.1 | 24 | 3415 | AAH21477.1 | |
BC034682.1 | 24 | 3359 | AAH34682.1 |