Assay Details
Target Gene Details
Entrez Gene ID: | 171580 |
Gene Name: | microtubule associated monooxygenase, calponin and LIM domain containing 1 |
Gene Aliases: |
MICAL, MICAL-1, Nical |
Location: |
Chr.10:41475860-41487030 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mical1 | NM_001164433.1 | 2 | 278 | NP_001157905.1 |
NM_138315.2 | 2 | 278 | NP_612188.1 | |
AK154665.1 | 2 | 218 | ||
AK154930.1 | 2 | 258 | ||
AK157196.1 | 2 | 217 | ||
AK159369.1 | 2 | 225 | BAE35027.1 | |
AK170382.1 | 2 | 217 | ||
AK170827.1 | 2 | 249 | ||
AK170874.1 | 2 | 260 | ||
AK170881.1 | 2 | 244 | BAE42090.1 | |
AK171234.1 | 2 | 245 | BAE42330.1 | |
AK171411.1 | 2 | 279 | BAE42437.1 | |
AK171429.1 | 2 | 257 | BAE42447.1 | |
AK171479.1 | 2 | 251 | BAE42481.1 | |
AK171779.1 | 2 | 257 | ||
AK172562.1 | 2 | 251 | ||
AK172716.1 | 2 | 204 | ||
AK178141.1 | ||||
AK184138.1 | ||||
AK190615.1 | ||||
AK206250.1 | ||||
AK218633.1 | ||||
BC021477.1 | 2 | 226 | AAH21477.1 | |
BC034682.1 | 2 | 170 | AAH34682.1 |