Assay Details
Target Gene Details
Entrez Gene ID: | 103220 |
Gene Name: | tetratricopeptide repeat domain 41 |
Gene Aliases: |
AI449705, AU041509, Gnn |
Location: |
Chr.10:86705632-86776844 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc41 | NM_001003910.2 | 16 | 4134 | NP_001003910.2 |
AK133195.1 | 16 | 4079 | ||
AK166553.1 | 8 | 1450 | BAE38851.1 | |
AY651021.1 | 15 | 3774 | AAT81233.1 | |
BG072108.2 | 1 | 232 |