Assay Details
Target Gene Details
Entrez Gene ID: | 20393 |
Gene Name: | serum/glucocorticoid regulated kinase 1 |
Gene Aliases: |
Sgk |
Location: |
Chr.10:21882184-21999903 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sgk1 | NM_001161845.2 | 1 | 350 | NP_001155317.2 |
AK043284.1 | 1 | 166 | ||
BC070401.1 | 1 | 350 | AAH70401.1 |
More Information
Set Membership: |
Intragenic Exonic |