Assay Details
Target Gene Details
Entrez Gene ID: | 71687 |
Gene Name: | transmembrane protein 25 |
Gene Aliases: |
0610039J01Rik, AI429491 |
Location: |
Chr.9:44793772-44799273 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem25 | NM_027865.2 | 9 | 1725 | NP_082141.1 |
AK002841.1 | 9 | 1726 | BAB22398.1 | |
AK090375.1 | 7 | 1999 | BAC41190.1 | |
AK154206.1 | 8 | 2211 | BAE32437.1 | |
AK165199.1 | 9 | 1757 | BAE38074.1 | |
AK165827.1 | 9 | 1710 | BAE38398.1 | |
BC060057.1 | 7 | 2048 | AAH60057.1 | |
BC060243.1 | 7 | 2048 | AAH60243.1 |