Assay Details
Target Gene Details
Entrez Gene ID: | 56351 |
Gene Name: | prostaglandin E synthase 3 (cytosolic) |
Gene Aliases: |
5730442A20Rik, Ptges, Tebp, cPGES, p23, sid3177 |
Location: |
Chr.10:128058982-128077254 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptges3 | NM_019766.4 | 8 | 1753 | NP_062740.1 |
AK077538.1 | 8 | 1754 | BAC36854.1 | |
AK088321.1 | 1 | 834 | ||
BC085264.1 | 8 | 1650 | AAH85264.1 |
More Information
Set Membership: |
Intragenic Exonic |