Assay Details
Target Gene Details
Entrez Gene ID: | 14421 |
Gene Name: | beta-1,4-N-acetyl-galactosaminyl transferase 1 |
Gene Aliases: |
4933429D13Rik, Gal-NAc-T, GalNAc-T, GalNAcT, Galgt1, Ggm-2, Ggm2 |
Location: |
Chr.10:127165156-127172340 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
B4galnt1 | NM_001244617.1 | 1 | 220 | NP_001231546.1 |
NM_008080.5 | 1 | 220 | NP_032106.1 | |
NR_045166.1 | 1 | 220 | ||
AK144484.1 | 1 | 186 | BAE25913.1 | |
BC022180.1 | AAH22180.1 | |||
BC057199.1 | 1 | 151 | AAH57199.1 |