Assay Details
Target Gene Details
Entrez Gene ID: | 56214 |
Gene Name: | secretory carrier membrane protein 4 |
Gene Aliases: |
2410022D05Rik, AI448940, AU022392, AW742369, Sc4 |
Location: |
Chr.10:80602882-80615783 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Scamp4 | NM_019575.4 | 7 | 1202 | NP_062521.1 |
AF224721.1 | 7 | 1177 | AAF35368.1 | |
AK019112.1 | 7 | 1205 | ||
AK154094.1 | 7 | 1193 | BAE32371.1 | |
AK160755.1 | 7 | 1193 | BAE35991.1 | |
AK168554.1 | 7 | 1188 | BAE40428.1 | |
BC018215.1 | 7 | 1171 | AAH18215.1 | |
BC054104.2 | 7 | 1197 | AAH54104.1 |
More Information
Set Membership: |
Intragenic Exonic |