Assay Details
Target Gene Details
Entrez Gene ID: | 53404 |
Gene Name: | atonal homolog 7 (Drosophila) |
Gene Aliases: |
Math5, bHLHa13 |
Location: |
Chr.10:63098020-63101284 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atoh7 | NM_016864.1 | 1 | 374 | NP_058560.1 |
AF071223.1 | 1 | 374 | AAC68868.1 | |
AK005214.1 | ||||
BC092234.1 | 1 | 727 |
More Information
Set Membership: |
Intragenic Exonic |