Assay Details
Target Gene Details
Entrez Gene ID: | 20224 |
Gene Name: | SAR1 gene homolog A (S. cerevisiae) |
Gene Aliases: |
1600019H17Rik, Sara, Sara1 |
Location: |
Chr.10:61680304-61693297 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sar1a | NM_009120.2 | 7 | 1589 | NP_033146.1 |
AK005495.1 | ||||
AK157843.1 | 7 | 1601 | BAE34224.1 | |
AK159239.1 | 7 | 1600 | BAE34922.1 | |
BC005549.1 | 7 | 1590 | AAH05549.1 |