Assay Details
Target Gene Details
Entrez Gene ID: | 20203 |
Gene Name: | S100 protein, beta polypeptide, neural |
Gene Aliases: |
AI850290, Bpb |
Location: |
Chr.10:76253836-76261319 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
S100b | NM_009115.3 | 3 | 1205 | NP_033141.1 |
AK134625.1 | 3 | 1132 | BAE22214.1 | |
AK135083.1 | 3 | 1188 | BAE22413.1 | |
AK161945.1 | 3 | 1181 | BAE36647.1 | |
AW456919.1 |
More Information
Set Membership: |
Intragenic Exonic |