Assay Details
Target Gene Details
Entrez Gene ID: | 117149 |
Gene Name: | toll-interleukin 1 receptor (TIR) domain-containing adaptor protein |
Gene Aliases: |
AA407980, C130027E04Rik, Mal, Tlr4ap, Wyatt |
Location: |
Chr.9:35184391-35200291 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tirap | NM_001177845.1 | 6 | 1107 | NP_001171316.1 |
NM_001177846.1 | 6 | 1089 | NP_001171317.1 | |
NM_001177847.1 | 6 | 1035 | NP_001171318.1 | |
NM_054096.2 | 6 | 1099 | NP_473437.2 | |
AF378130.1 | 6 | 1008 | AAL05628.1 | |
AF410784.1 | 5 | 931 | AAL05037.1 | |
AK036431.1 | 6 | 1028 | BAC29426.1 | |
AK080135.1 | 6 | 1108 | BAC37831.1 | |
AK151117.1 | 6 | 1037 | BAE30126.1 | |
AK168494.1 | 6 | 1027 | BAE40381.1 | |
BC005584.1 | 3 | 765 | AAH05584.1 |