Assay Details
Target Gene Details
Entrez Gene ID: | 14775 |
Gene Name: | glutathione peroxidase 1 |
Gene Aliases: |
AI195024, AL033363, CGPx, GPx-1, GSHPx-1, Gpx |
Location: |
Chr.9:108339080-108340342 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpx1 | NM_008160.6 | 1 | 379 | NP_032186.2 |
AK002245.1 | 1 | 184 | BAC55244.1 | |
AK010999.1 | 1 | 184 | BAC55252.1 | |
AK011019.1 | 1 | 184 | BAC55253.1 | |
AK028171.1 | 1 | 184 | BAC55257.1 | |
AK150548.1 | 1 | 185 | BAE29650.1 | |
AK154833.1 | 1 | 184 | BAE32862.1 | |
AK160388.1 | 1 | 185 | BAE35760.1 | |
AK191208.1 | ||||
AK191913.1 | ||||
AK200043.1 | ||||
AK201721.1 | ||||
AK202661.1 | ||||
AK205185.1 | ||||
BC086649.1 | 1 | 167 | AAH86649.1 | |
CK628430.1 | 1 | 379 |
More Information
Set Membership: |
Intragenic Exonic |