Assay Details
Target Gene Details
Entrez Gene ID: | 73106 |
Gene Name: | protease, serine 57 |
Gene Aliases: |
2900092M14Rik, GLGL782, Prssl1, UNQ782 |
Location: |
Chr.10:79781474-79795605 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss57 | NM_001042710.1 | 4 | 663 | NP_001036175.1 |
AK013841.1 | 1 | 178 | ||
AK153740.1 | 4 | 630 | ||
AK158376.1 | 4 | 663 | ||
BC116380.2 | 5 | 813 |
More Information
Set Membership: |
Intragenic Exonic |