Assay Details
Target Gene Details
Entrez Gene ID: | 208084 |
Gene Name: | PIF1 5'-to-3' DNA helicase homolog (S. cerevisiae) |
Gene Aliases: |
4631410M14, AI449441 |
Location: |
Chr.9:65587147-65595967 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pif1 | NM_172453.3 | 1 | 796 | NP_766041.1 |
AK028460.1 | 1 | 797 | BAC25963.1 | |
AK049353.1 | 1 | 798 | BAC33702.1 | |
AK154072.1 | 1 | 809 | ||
AY100322.1 | 2 | 509 | AAM50051.1 | |
AY498715.1 | 1 | 796 | AAS77397.1 | |
BC046611.1 | 2 | 420 | AAH46611.1 |
More Information
Set Membership: |
Intragenic Exonic |