Assay Details
Target Gene Details
Entrez Gene ID: | 64011 |
Gene Name: | neurogranin |
Gene Aliases: |
0710001B06Rik, AI838505, NG, NG/RC3, Pss1, R75334, RC3 |
Location: |
Chr.9:37544493-37552745 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrgn | NM_022029.2 | 4 | 1012 | NP_071312.1 |
AK158630.1 | 4 | 999 | BAE34589.1 |
Target Gene Details
Entrez Gene ID: | 57276 |
Gene Name: | V-set and immunoglobulin domain containing 2 |
Gene Aliases: |
1190004B15Rik, 2210413P10Rik, AU040484, CTX, Ctm |
Location: |
Chr.9:37538814-37545123 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vsig2 |