Assay Details
Target Gene Details
Entrez Gene ID: | 68652 |
Gene Name: | TGF-beta activated kinase 1/MAP3K7 binding protein 2 |
Gene Aliases: |
1110030N06Rik, A530078N03Rik, Map3k7ip2, mKIAA0733 |
Location: |
Chr.10:7905648-7956225 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tab2 | NM_138667.3 | 7 | 2992 | NP_619608.1 |
AB093262.1 | 6 | 2559 | BAC41446.1 | |
AK031864.1 | 1 | 52 | ||
BC004072.1 | 5 | 1141 | AAH04072.1 | |
BC004813.1 | 6 | 2659 | AAH04813.1 | |
BF468714.1 | 1 | 128 |