Assay Details
Target Gene Details
Entrez Gene ID: | 68652 |
Gene Name: | TGF-beta activated kinase 1/MAP3K7 binding protein 2 |
Gene Aliases: |
1110030N06Rik, A530078N03Rik, Map3k7ip2, mKIAA0733 |
Location: |
Chr.10:7905648-7956225 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tab2 | NM_138667.3 | 3 | 1012 | NP_619608.1 |
AB093262.1 | 2 | 579 | BAC41446.1 | |
AK041067.1 | 2 | 1537 | ||
AK089164.1 | 3 | 1012 | BAC40772.1 | |
AK147830.1 | 3 | 823 | BAE28166.1 | |
AY093701.1 | 2 | 672 | AAM10487.1 | |
BC004813.1 | 2 | 679 | AAH04813.1 |