Assay Details
Target Gene Details
Entrez Gene ID: | 14360 |
Gene Name: | Fyn proto-oncogene |
Gene Aliases: |
AI448320, AW552119 |
Location: |
Chr.10:39369770-39565381 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fyn | NM_001122892.1 | 1 | 124 | NP_001116364.1 |
NM_001122893.1 | 1 | 124 | NP_001116365.1 | |
NM_008054.2 | 1 | 124 | NP_032080.2 | |
AK156584.1 | 1 | 126 | BAE33766.1 | |
AK171646.1 | 1 | 126 | BAE42585.1 | |
BC092217.1 | 1 | 120 | AAH92217.1 |