Assay Details
Target Gene Details
Entrez Gene ID: | 66706 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 3 |
Gene Aliases: |
4733401H18Rik |
Location: |
Chr.9:108565865-108567342 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufaf3 | NM_023247.1 | 1 | 416 | NP_075736.1 |
AF235017.1 | 1 | 416 | AAF61723.1 | |
AK005252.1 | BAB23907.1 | |||
AK014634.1 | BAB29477.1 | |||
BC027513.1 | 1 | 449 | AAH27513.1 |