Assay Details
Target Gene Details
Entrez Gene ID: | 19886 |
Gene Name: | Ros1 proto-oncogene |
Gene Aliases: |
Ros-1, c-ros |
Location: |
Chr.10:52044865-52195397 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ros1 | NM_011282.2 | 22 | 3621 | NP_035412.2 |
U15443.1 | 22 | 3620 | AAA50600.1 | |
X81650.1 | 22 | 3444 | CAA57310.1 |
More Information
Set Membership: |
Intragenic Exonic |