Assay Details
Target Gene Details
Entrez Gene ID: | 71764 |
Gene Name: | C2 calcium-dependent domain containing 2-like |
Gene Aliases: |
1300006O23Rik, Tmem24 |
Location: |
Chr.9:44309237-44320324 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C2cd2l | NM_027909.2 | 13 | 2260 | NP_082185.2 |
AK004920.1 | 13 | 2265 | BAB23671.1 | |
AK039220.1 | 13 | 2231 | BAC30281.1 | |
AK122237.1 | 8 | 3835 | BAC65519.1 | |
AK153804.1 | 13 | 2232 | BAE32190.1 | |
BC035271.1 | 10 | 2266 | AAH35271.1 | |
BC049875.1 | 9 | 1187 | AAH49875.1 | |
BC056612.1 | 13 | 1831 | AAH56612.1 | |
BC060156.1 | 13 | 2124 | AAH60156.1 |