Assay Details
Target Gene Details
Entrez Gene ID: | 170732 |
Gene Name: | thyrotropin releasing hormone receptor 2 |
Gene Aliases: |
TRH-R2 |
Location: |
Chr.8:122356065-122362910 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trhr2 | NM_133202.2 | 2 | 480 | NP_573465.1 |
AF283762.1 | 2 | 480 | AAG17712.1 | |
BC117988.2 | 2 | 459 |
More Information
Set Membership: |
Intragenic Exonic |