Assay Details
Target Gene Details
Entrez Gene ID: | 234374 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 49 |
Gene Aliases: |
R27090_2 |
Location: |
Chr.8:70292866-70302511 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx49 | NM_001024922.2 | 13 | 1695 | NP_001020093.2 |
Target Gene Details
Entrez Gene ID: | 26558 |
Gene Name: | homer homolog 3 (Drosophila) |
Gene Aliases: |
AW146114 |
Location: |
Chr.8:70282891-70294361 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Homer3 | NM_001146153.1 | 10 | 1355 | NP_001139625.1 |
NM_011984.2 | 10 | 1346 | NP_036114.2 | |
AF093261.1 | 8 | 1182 | AAC71025.1 | |
AK203312.1 | ||||
AK215706.1 | ||||
AK217189.1 | ||||
BC005773.1 | 10 | 1295 | AAH05773.1 |