Assay Details
Target Gene Details
Entrez Gene ID: | 20295 |
Gene Name: | chemokine (C-C motif) ligand 17 |
Gene Aliases: |
Abcd-2, Scya17, Scya17l, Tarc |
Location: |
Chr.8:94807404-94812038 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl17 | NM_011332.3 | 3 | 419 | NP_035462.2 |
AF125570.1 | 3 | 508 | AAD56600.1 | |
AF125571.1 | 3 | 450 | AAD56601.1 | |
AF125572.1 | 3 | 423 | AAD56602.1 | |
AJ242587.1 | 3 | 398 | CAB45256.1 | |
AK007663.1 | 3 | 420 | BAB25171.1 | |
BC028505.1 | 3 | 403 | AAH28505.1 |
More Information
Set Membership: |
Intragenic Exonic |