Assay Details
Target Gene Details
Entrez Gene ID: | 113868 |
Gene Name: | acetyl-Coenzyme A acyltransferase 1A |
Gene Aliases: |
Acaa, Acaa1, D9Ertd25e, PTL |
Location: |
Chr.9:119341052-119350296 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acaa1a | NM_130864.3 | 1 | 61 | NP_570934.1 |
AY273811.1 | 1 | 61 | AAP31668.1 |
More Information
Set Membership: |
Intragenic Exonic |