Assay Details
Target Gene Details
Entrez Gene ID: | 18168 |
Gene Name: | neuropeptide Y receptor Y5 |
Gene Aliases: |
NPY5-R, NPYY5-R, Y5R |
Location: |
Chr.8:66679965-66688581 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npy5r | NM_016708.3 | 2 | 498 | NP_057917.2 |
AB001346.1 | 2 | 501 | BAA89538.1 | |
AF049329.1 | 1 | 369 | AAC17839.1 | |
AK045587.1 | 2 | 447 | BAC32426.1 | |
BC116976.1 | 2 | 451 | ||
BC116978.1 | 2 | 535 |
More Information
Set Membership: |
Intragenic Exonic |