Assay Details
Target Gene Details
Entrez Gene ID: | 101966 |
Gene Name: | DNA segment, Chr 8, ERATO Doi 738, expressed |
Gene Aliases: |
AA589584 |
Location: |
Chr.8:84246235-84249761 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
D8Ertd738e | NM_001007571.2 | 3 | 500 | NP_001007572.1 |
AK146163.1 | 3 | 458 | BAE26944.1 | |
AK167785.1 | 3 | 459 | BAE39817.1 | |
AK215593.1 | 1 | 260 | ||
BC024662.1 | 3 | 449 | AAH24662.1 | |
BF465076.1 | 1 | 184 | ||
CA318976.1 | 3 | 440 | ||
CF740495.1 | 3 | 500 |