Assay Details
Target Gene Details
Entrez Gene ID: | 29873 |
Gene Name: | chondroitin sulfate proteoglycan 5 |
Gene Aliases: |
Caleb, Ngc |
Location: |
Chr.9:110243783-110262576 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cspg5 | NM_001166273.1 | 5 | 3442 | NP_001159745.1 |
NM_013884.3 | 5 | 3361 | NP_038912.3 | |
AF133700.1 | 5 | 1546 | AAF23362.1 | |
AF461090.1 | 5 | 3355 | AAQ04777.1 | |
AF461091.1 | 4 | 1679 | AAQ04778.1 | |
AF461092.1 | 5 | 3436 | AAQ04779.1 | |
AK031799.1 | 1 | 1788 | ||
BC055736.1 | 5 | 1693 | AAH55736.1 |