Assay Details
Target Gene Details
Entrez Gene ID: | 16598 |
Gene Name: | Kruppel-like factor 2 (lung) |
Gene Aliases: |
Lklf |
Location: |
Chr.8:72319062-72321654 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klf2 | NM_008452.2 | 2 | 224 | NP_032478.2 |
AK131962.1 | 2 | 225 | BAE20905.1 | |
BC138737.1 | 2 | 232 | ||
BC138738.1 | 2 | 232 | ||
BF023555.1 | 2 | 206 | ||
U25096.1 | 2 | 253 | AAA86728.1 |
More Information
Set Membership: |
Intragenic Exonic |