Assay Details
Target Gene Details
Entrez Gene ID: | 67213 |
Gene Name: | CKLF-like MARVEL transmembrane domain containing 6 |
Gene Aliases: |
2810051A14Rik, AA536733, Cklfsf6 |
Location: |
Chr.9:114731157-114749347 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cmtm6 | NM_026036.3 | 4 | 921 | NP_080312.1 |
AK012941.1 | 4 | 922 | BAB28562.1 | |
AK030885.1 | 4 | 901 | BAC27172.1 | |
AK128960.1 | 4 | 907 | ||
AK145743.1 | 4 | 902 | BAE26621.1 | |
AK150775.1 | 4 | 920 | ||
AK151092.1 | 4 | 920 | ||
AK151249.1 | 1 | 440 | BAE30239.1 | |
AK151431.1 | 4 | 920 | ||
AK152448.1 | 4 | 923 | BAE31227.1 | |
AK152723.1 | 4 | 932 | BAE31446.1 | |
AK152817.1 | 4 | 932 | BAE31520.1 | |
AK153092.1 | 4 | 920 | ||
AK154481.1 | 4 | 908 | BAE32617.1 | |
AK159439.1 | 4 | 891 | BAE35085.1 | |
AK159735.1 | 4 | 916 | BAE35329.1 | |
AK165948.1 | 4 | 890 | BAE38480.1 | |
AK167499.1 | 4 | 908 | BAE39575.1 | |
AK167854.1 | 4 | 913 | BAE39872.1 | |
AK170905.1 | 4 | 923 | BAE42106.1 | |
AK171394.1 | 4 | 952 | BAE42431.1 | |
AK204614.1 | ||||
AK205076.1 | ||||
AY241868.1 | 4 | 958 | AAP33490.1 | |
BC007194.1 | 2 | 569 | AAH07194.1 | |
BC027248.1 | 4 | 893 | AAH27248.1 |
More Information
Set Membership: |
Intragenic Exonic |