Assay Details
Target Gene Details
Entrez Gene ID: | 18997 |
Gene Name: | POU domain, class 4, transcription factor 2 |
Gene Aliases: |
Brn-3.2, Brn-3b, Brn3b, Pou4f-rs1, mBrn3-3R |
Location: |
Chr.8:78433009-78436652 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pou4f2 | NM_138944.2 | 1 | 87 | NP_620394.2 |
AK084240.1 | 1 | 83 | ||
AK087546.1 | 1 | 87 | BAC39921.1 | |
S68377.1 | 1 | 69 | AAC60672.1 |