Assay Details
Target Gene Details
Entrez Gene ID: | 11945 |
Gene Name: | ATPase, H+/K+ exchanging, beta polypeptide |
Gene Aliases: |
AV080843 |
Location: |
Chr.8:13386209-13396778 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp4b | NM_009724.2 | 7 | 1233 | NP_033854.1 |
AK008180.1 | 7 | 1232 | BAB25514.1 | |
AK009499.1 | 7 | 1233 | BAB26326.1 | |
AK009670.1 | 7 | 1237 | BAC25265.1 | |
AK009732.1 | 1 | 330 | ||
AK009832.1 | 7 | 1235 | BAB26530.1 | |
AK009994.1 | 7 | 1232 | BAB26632.1 | |
AK010003.1 | 7 | 1232 | BAB26635.1 | |
AK010030.1 | 7 | 1233 | BAB26654.1 | |
AK010049.1 | 7 | 1280 | BAB26665.1 |
More Information
Set Membership: |
Intragenic Exonic |