Assay Details
Target Gene Details
Entrez Gene ID: | 233905 |
Gene Name: | zinc finger protein 646 |
Gene Aliases: |
6820429M01 |
Location: |
Chr.7:127877036-127885996 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp646 | NM_172749.4 | 2 | 3082 | NP_766337.2 |
AK078520.1 | 2 | 2805 | BAC37321.1 | |
AK129112.1 | 1 | 1934 | BAC97922.1 | |
BC068300.1 | 1 | 2685 | AAH68300.1 | |
BC079907.1 | 2 | 3082 | AAH79907.1 |
More Information
Set Membership: |
Intragenic Exonic |